3-Bromo-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

23642-5G 23642-25G 23642-1G 23642-500MG
77628-196EA 1484.23 USD
77628-196 77635-550 77637-386 77645-378
3-Bromo-2-(methylthio)benzoic acid ≥97%
3-Bromo-2-(methylthio)benzoic acid

Precaución: For Research Use Only

Formula: 247.109 g/mol
MW: C₈H₇BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD16036855
CAS Number: 503821-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR