5-Bromo-4-methyl-3-pyridinecarboxamide ≥97%

Supplier: AOBChem USA

Warning

23633-5G 23633-25G 23633-1G 23633-500MG
77632-160EA 719 USD
77632-160 77654-336 77641-398 77649-414
5-Bromo-4-methyl-3-pyridinecarboxamide ≥97%
5-Bromo-4-methyl-3-pyridinecarboxamide

Precaución: For Research Use Only

Formula: 215.047 g/mol
MW: C₇H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD07439980
CAS Number: 677702-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR