5-Bromo-4-methyl-3,3'-bipyridine ≥97%

Supplier: AOBChem USA

Warning

23632-5G 23632-1G 23632-25G 23632-500MG
77632-158EA 1302 USD
77632-158 77637-384 77658-144 77645-376
5-Bromo-4-methyl-3,3'-bipyridine ≥97%
5-Bromo-4-methyl-3,3'-bipyridine

Precaución: For Research Use Only

Formula: 249.107 g/mol
MW: C₁₁H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD22543665
CAS Number: 351457-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR