4-(5-Bromo-2-pyridinyl)phenol ≥97%

Supplier: AOBChem USA

Warning

23617-5G 23617-1G 23617-25G 23617-500MG
77632-156EA 1430 USD
77632-156 77637-382 77658-142 77645-374
4-(5-Bromo-2-pyridinyl)phenol ≥97%
4-(5-Bromo-2-pyridinyl)phenol

Precaución: For Research Use Only

Formula: 250.091 g/mol
MW: C₁₁H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD28339814
CAS Number: 1032825-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR