6-(4-Fluorophenyl)nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

23610-5G 23610-25G 23610-1G 23610-500MG
77632-152EA 903 USD
77632-152 77654-334 77641-396 77649-412
6-(4-Fluorophenyl)nicotinaldehyde ≥97%
6-(4-Fluorophenyl)nicotinaldehyde

Precaución: For Research Use Only

Formula: C₁₂H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD04115445
CAS Number: 135958-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR