α-Ethenyl-4-methoxy-3,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235218-500MG 235218-5G 235218-1G
77652-528EA 384 USD
77652-528 77658-138 77641-394
α-Ethenyl-4-methoxy-3,5-dimethylbenzenemethanol ≥95%
α-Ethenyl-4-methoxy-3,5-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD26621821
CAS Number: 1541276-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR