α-Ethenyl-5-fluoro-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235184-5G 235184-1G 235184-500MG
77628-192EA 1484.23 USD
77628-192 77637-376 77645-368
α-Ethenyl-5-fluoro-2-methylbenzenemethanol ≥95%
α-Ethenyl-5-fluoro-2-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD34870256
CAS Number: 1602219-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR