3-Bromo-α-ethenyl-2-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235173-5G 235173-1G 235173-500MG
77628-190EA 1703.62 USD
77628-190 77637-374 77645-366
3-Bromo-α-ethenyl-2-fluorobenzenemethanol ≥95%
3-Bromo-α-ethenyl-2-fluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₉H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD34870248
CAS Number: 149946-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR