3-Iodo-2-piperidin-1-ylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
23514-5G
23514-500MG
23514-25G
23514-1G
77632-148EA
639
USD
77632-148
77652-526
77654-330
77641-390
3-Iodo-2-piperidin-1-ylpyridine ≥97%
3-Iodo-2-piperidin-1-ylpyridine
Precaución: For Research Use Only
Formula:
C₁₀H₁₃IN₂ Storage Temperature: Ambient |
MDL Number:
MFCD10703541 CAS Number: 1086376-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...