α-Ethenyl-3-fluoro-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235102-1G 235102-5G 235102-500MG
77637-370EA 700 USD
77637-370 77654-328 77645-364
α-Ethenyl-3-fluoro-2-methylbenzenemethanol ≥95%
α-Ethenyl-3-fluoro-2-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD34870236
CAS Number: 1936638-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR