3-Chloro-α-ethenyl-2-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

235080-5G 235080-1G 235080-500MG
77628-184EA 1484.23 USD
77628-184 77641-386 77649-404
3-Chloro-α-ethenyl-2-fluorobenzenemethanol ≥97%
3-Chloro-α-ethenyl-2-fluorobenzenemethanol

Precaución: For Research Use Only

Formula: 186.61 g/mol
MW: C₉H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28153824
CAS Number: 149946-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR