α-Ethenyl-4-fluoro-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235076-5G 235076-500MG 235076-1G
77628-182EA 1484.23 USD
77628-182 77652-524 77641-384
α-Ethenyl-4-fluoro-2-methylbenzenemethanol ≥95%
α-Ethenyl-4-fluoro-2-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD21232158
CAS Number: 1430748-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR