α-Ethenyl-2-methoxy-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235042-5G 235042-25G 235042-1G 235042-500MG
77628-180EA 1484.23 USD
77628-180 77635-546 77641-382 77645-362
α-Ethenyl-2-methoxy-3-methylbenzenemethanol ≥95%
α-Ethenyl-2-methoxy-3-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34870225
CAS Number: 2228723-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR