α-Ethenyl-3-methoxy-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235024-5G 235024-25G 235024-1G 235024-500MG
77628-178EA 1484.23 USD
77628-178 77635-544 77637-366 77645-360
α-Ethenyl-3-methoxy-4-methylbenzenemethanol ≥95%
α-Ethenyl-3-methoxy-4-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34870218
CAS Number: 1595049-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR