α-Ethenyl-5-methoxy-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235014-5G 235014-1G 235014-500MG
77654-324EA 2417 USD
77654-324 77641-380 77649-398
α-Ethenyl-5-methoxy-2-methylbenzenemethanol ≥95%
α-Ethenyl-5-methoxy-2-methylbenzenemethanol

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34870215
CAS Number: 2294964-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR