3,5-Dichloro-α-ethenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235000-5G 235000-1G 235000-25G 235000-500MG
77628-174EA 1720.1 USD
77628-174 77637-362 77654-322 77645-358
3,5-Dichloro-α-ethenylbenzenemethanol ≥95%
3,5-Dichloro-α-ethenylbenzenemethanol

Precaución: For Research Use Only

Formula: 203.07 g/mol
MW: C₉H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD21232160
CAS Number: 149946-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR