α-Ethenyl-2,4-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234936-5G 234936-1G 234936-25G 234936-500MG
77632-144EA 1320 USD
77632-144 77637-360 77658-134 77645-350
α-Ethenyl-2,4-dimethylbenzenemethanol ≥95%
α-Ethenyl-2,4-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD21232124
CAS Number: 860590-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR