4-(1,1-Dimethylethyl)-α-ethenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234913-5G 234913-25G 234913-1G 234913-500MG
77632-142EA 1320 USD
77632-142 77658-132 77641-376 77645-348
4-(1,1-Dimethylethyl)-α-ethenylbenzenemethanol ≥95%
4-(1,1-Dimethylethyl)-α-ethenylbenzenemethanol

Precaución: For Research Use Only

Formula: 190.28 g/mol
MW: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD21232085
CAS Number: 863029-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR