α-Ethenyl-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234906-5G 234906-1G 234906-500MG
77658-130EA 2608 USD
77658-130 77641-374 77649-396
α-Ethenyl-2-methoxybenzenemethanol ≥95%
α-Ethenyl-2-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD21232060
CAS Number: 58824-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR