α-Ethenyl-3-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234905-5G 234905-25G 234905-1G 234905-500MG
77628-168EA 1553.24 USD
77628-168 77635-540 77641-372 77645-346
α-Ethenyl-3-methoxybenzenemethanol ≥95%
α-Ethenyl-3-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD21232064
CAS Number: 58824-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR