α-Ethenyl-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

234904-5G 234904-1G 234904-500MG 234904-25G
77632-140EA 1320 USD
77632-140 77637-358 77652-522 77654-320
α-Ethenyl-1-naphthalenemethanol ≥95%
α-Ethenyl-1-naphthalenemethanol

Precaución: For Research Use Only

Formula: C₁₃H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD21232098
CAS Number: 61619-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR