α-Ethenyl-2-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

234901-5G 234901-1G 234901-25G 234901-500MG
77632-138EA 1320 USD
77632-138 77637-356 77658-128 77645-344
α-Ethenyl-2-naphthalenemethanol ≥95%
α-Ethenyl-2-naphthalenemethanol

Precaución: For Research Use Only

Formula: C₁₃H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD21232127
CAS Number: 76635-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR