4-Bromo-α-ethenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234900-1G 234900-5G 234900-500MG
77634-448EA 794 USD
77634-448 77654-318 77645-342
4-Bromo-α-ethenylbenzenemethanol ≥95%
4-Bromo-α-ethenylbenzenemethanol

Precaución: For Research Use Only

Formula: 213.07 g/mol
MW: C₉H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD21232082
CAS Number: 58824-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR