2,5-Difluoro-α-methyl-α-phenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234890-5G 234890-25G 234890-1G 234890-500MG
77628-166EA 1484.23 USD
77628-166 77658-126 77641-370 77649-394
2,5-Difluoro-α-methyl-α-phenylbenzenemethanol ≥95%
2,5-Difluoro-α-methyl-α-phenylbenzenemethanol

Precaución: For Research Use Only

Formula: 234.24 g/mol
MW: C₁₄H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD18023591
CAS Number: 1281647-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR