4-Fluoro-α-methyl-α-phenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234755-5G 234755-1G 234755-500MG
77654-316EA 1900 USD
77654-316 77641-368 77649-392
4-Fluoro-α-methyl-α-phenylbenzenemethanol ≥95%
4-Fluoro-α-methyl-α-phenylbenzenemethanol

Precaución: For Research Use Only

Formula: 216.25 g/mol
MW: C₁₄H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD12067983
CAS Number: 339-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR