2-Bromo-3,5-dimethylbenzenamine ≥97%

Supplier: AOBChem USA

Warning

234615-500MG 234615-5G 234615-1G
77652-520EA 392 USD
77652-520 77658-124 77641-366
2-Bromo-3,5-dimethylbenzenamine ≥97%
2-Bromo-3,5-dimethylbenzenamine

Precaución: For Research Use Only

Formula: 200.08 g/mol
MW: C₈H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD24623799
CAS Number: 35490-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR