Benzophenone, 4-chloro-3-fluoro- ≥95%

Supplier: AOBChem USA

Warning

234556-5G 234556-25G 234556-1G 234556-500MG
77632-136EA 1381 USD
77632-136 77658-122 77641-362 77645-340
Benzophenone, 4-chloro-3-fluoro- ≥95%
Benzophenone, 4-chloro-3-fluoro-

Precaución: For Research Use Only

Formula: C₁₃H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD20920411
CAS Number: 16197-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR