1-(3-Methoxy-4-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

234464-5G 234464-25G 234464-1G 234464-500MG
77628-164EA 1484.23 USD
77628-164 77658-118 77641-360 77649-388
1-(3-Methoxy-4-methylphenyl)-1-propanone ≥95%
1-(3-Methoxy-4-methylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD24695156
CAS Number: 18158-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR