(3,5-Dichlorophenyl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

234429-5G 234429-1G 234429-500MG
77654-310EA 2001 USD
77654-310 77641-358 77645-338
(3,5-Dichlorophenyl)phenylmethanone ≥95%
(3,5-Dichlorophenyl)phenylmethanone

Precaución: For Research Use Only

Formula: 251.11 g/mol
MW: C₁₃H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD00045188
CAS Number: 13395-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR