Cyclopropyl(3-ethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

234419-5G 234419-1G 234419-500MG
77654-308EA 2001 USD
77654-308 77641-356 77645-336
Cyclopropyl(3-ethylphenyl)methanone ≥95%
Cyclopropyl(3-ethylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD24127325
CAS Number: 1528975-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR