4-Iodo-2-(1-piperidinyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

23434-5G 23434-1G 23434-500MG 23434-25G
77634-444EA 288 USD
77634-444 77637-350 77652-514 77658-116
4-Iodo-2-(1-piperidinyl)pyridine ≥97%
4-Iodo-2-(1-piperidinyl)pyridine

Precaución: For Research Use Only

Formula: 288.128 g/mol
MW: C₁₀H₁₃IN₂
Storage Temperature: Ambient
MDL Number: MFCD22056767
CAS Number: 1239969-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR