(3-Chloro-4-methoxyphenyl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

234275-5G 234275-25G 234275-1G 234275-500MG
77632-134EA 1441 USD
77632-134 77654-304 77641-352 77649-384
(3-Chloro-4-methoxyphenyl)phenylmethanone ≥95%
(3-Chloro-4-methoxyphenyl)phenylmethanone

Precaución: For Research Use Only

Formula: 246.69 g/mol
MW: C₁₄H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD01464001
CAS Number: 10547-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR