1-(3-Ethoxy-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

234206-5G 234206-1G 234206-25G 234206-500MG
77632-132EA 1180 USD
77632-132 77637-348 77658-114 77645-334
1-(3-Ethoxy-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Ethoxy-4-methylphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18375524
CAS Number: 1443312-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR