2-Methyl-1-[3-(1-methylethyl)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

234205-1G 234205-5G 234205-500MG
77637-346EA 591 USD
77637-346 77658-112 77645-332
2-Methyl-1-[3-(1-methylethyl)phenyl]-1-propanone ≥95%
2-Methyl-1-[3-(1-methylethyl)phenyl]-1-propanone

Precaución: For Research Use Only

Formula: 190.28 g/mol
MW: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD22373032
CAS Number: 1443313-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR