4-Chloro-2,3-difluoro-5-methoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

23410-5G 23410-1G 23410-500MG
77628-162EA 1484.23 USD
77628-162 77637-344 77649-382
4-Chloro-2,3-difluoro-5-methoxyphenylboronic acid ≥95%
4-Chloro-2,3-difluoro-5-methoxyphenylboronic acid

Precaución: For Research Use Only

Formula: 222.382 g/mol
MW: C₇H₆BClF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30529798
CAS Number: 2121513-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR