4-Iodo-2-phenoxypyridine ≥97%

Supplier: AOBChem USA

Warning

23400-5G 23400-1G 23400-25G 23400-500MG
77634-442EA 534 USD
77634-442 77637-342 77658-110 77649-380
4-Iodo-2-phenoxypyridine ≥97%
4-Iodo-2-phenoxypyridine

Precaución: For Research Use Only

Formula: C₁₁H₈INO
Storage Temperature: Ambient
MDL Number: MFCD22056765
CAS Number: 1353776-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR