1-Propanone, 1-(3-chloro-4-methylphenyl)-2-methyl- ≥95%

Supplier: AOBChem USA

Warning

233861-5G 233861-25G 233861-1G 233861-500MG
77632-128EA 1159 USD
77632-128 77654-298 77641-350 77649-378
1-Propanone, 1-(3-chloro-4-methylphenyl)-2-methyl- ≥95%
1-Propanone, 1-(3-chloro-4-methylphenyl)-2-methyl-

Precaución: For Research Use Only

Formula: 196.67 g/mol
MW: C₁₁H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD19314976
CAS Number: 75187-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR