Cyclopropyl(3,4-dimethoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

233837-5G 233837-25G 233837-1G 233837-500MG
77632-126EA 1261 USD
77632-126 77654-296 77641-348 77649-376
Cyclopropyl(3,4-dimethoxyphenyl)methanone ≥95%
Cyclopropyl(3,4-dimethoxyphenyl)methanone

Precaución: For Research Use Only

Formula: 206.24 g/mol
MW: C₁₂H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD08685667
CAS Number: 92847-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR