2-Fluoro-N-methoxy-N-methylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

23379-5G 23379-1G 23379-500MG 23379-25G
77634-440EA 470 USD
77634-440 77637-338 77652-512 77658-108
2-Fluoro-N-methoxy-N-methylnicotinamide ≥97%
2-Fluoro-N-methoxy-N-methylnicotinamide

Precaución: For Research Use Only

Formula: 184.168 g/mol
MW: C₈H₉FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11847416
CAS Number: 949154-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR