Cyclohexyl-3-methoxyphenyl ketone ≥95%

Supplier: AOBChem USA

Warning

233512-5G 233512-1G 233512-500MG
77628-160EA 1484.23 USD
77628-160 77641-344 77645-324
Cyclohexyl-3-methoxyphenyl ketone ≥95%
Cyclohexyl-3-methoxyphenyl ketone

Precaución: For Research Use Only

Formula: C₁₄H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD03841340
CAS Number: 898792-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR