1-Propanone, 1-(3,5-difluorophenyl)-2-methyl- ≥95%

Supplier: AOBChem USA

Warning

233508-5G 233508-25G 233508-1G 233508-500MG
77632-122EA 1159 USD
77632-122 77654-290 77641-342 77645-322
1-Propanone, 1-(3,5-difluorophenyl)-2-methyl- ≥95%
1-Propanone, 1-(3,5-difluorophenyl)-2-methyl-

Precaución: For Research Use Only

Formula: 184.18 g/mol
MW: C₁₀H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD16069386
CAS Number: 1226351-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR