1-(3,5-Dichlorophenyl)-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

233428-5G 233428-25G 233428-1G 233428-500MG
77632-114EA 913 USD
77632-114 77658-102 77641-338 77645-318
1-(3,5-Dichlorophenyl)-2-methyl-1-propanone ≥95%
1-(3,5-Dichlorophenyl)-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: C₁₀H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD19317593
CAS Number: 1267546-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR