Methanone, cyclopropyl(3,5-dichlorophenyl)- ≥95%

Supplier: AOBChem USA

Warning

233418-5G 233418-25G 233418-1G 233418-500MG
77632-112EA 1203 USD
77632-112 77658-100 77641-336 77645-316
Methanone, cyclopropyl(3,5-dichlorophenyl)- ≥95%
Methanone, cyclopropyl(3,5-dichlorophenyl)-

Precaución: For Research Use Only

Formula: C₁₀H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD03841240
CAS Number: 898790-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR