Cyclobutyl(3,4-dimethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

233407-5G 233407-1G 233407-500MG
77628-158EA 1703.62 USD
77628-158 77637-330 77652-510
Cyclobutyl(3,4-dimethylphenyl)methanone ≥95%
Cyclobutyl(3,4-dimethylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₃H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD03841268
CAS Number: 898790-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR