Cyclopentyl(3-fluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

233396-5G 233396-1G 233396-25G 233396-500MG
77632-110EA 1412 USD
77632-110 77637-326 77654-284 77645-312
Cyclopentyl(3-fluorophenyl)methanone ≥95%
Cyclopentyl(3-fluorophenyl)methanone

Precaución: For Research Use Only

Formula: 192.23 g/mol
MW: C₁₂H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD03841309
CAS Number: 898791-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR