1-(3-Chloro-4-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

233393-5G 233393-25G 233393-1G 233393-500MG
77632-108EA 998 USD
77632-108 77658-098 77641-334 77649-366
1-(3-Chloro-4-methylphenyl)-1-propanone ≥95%
1-(3-Chloro-4-methylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 182.65 g/mol
MW: C₁₀H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD11617660
CAS Number: 149758-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR