Cyclobutyl(3,4-dichlorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

233361-5G 233361-1G 233361-500MG
77628-154EA 1484.23 USD
77628-154 77641-332 77649-364
Cyclobutyl(3,4-dichlorophenyl)methanone ≥95%
Cyclobutyl(3,4-dichlorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₁H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD03841285
CAS Number: 898791-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR