1-(Benzyloxy)-4-bromo-2-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

23322-5G 23322-25G 23322-1G
77634-434EA 198 USD
77634-434 77658-096 77641-328
1-(Benzyloxy)-4-bromo-2-(trifluoromethoxy)benzene ≥95%
1-(Benzyloxy)-4-bromo-2-(trifluoromethoxy)benzene

Precaución: For Research Use Only

Formula: 347.127 g/mol
MW: C₁₄H₁₀BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34178489
CAS Number: 647856-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR