1-(3-Chloro-4-methoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

233209-5G 233209-25G 233209-1G 233209-500MG
77628-152EA 1484.23 USD
77628-152 77654-278 77641-324 77649-362
1-(3-Chloro-4-methoxyphenyl)propan-1-one ≥95%
1-(3-Chloro-4-methoxyphenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11199128
CAS Number: 4394-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR