Cyclopentyl(3,4-dimethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

233207-5G 233207-1G 233207-25G 233207-500MG
77634-430EA 391 USD
77634-430 77637-324 77654-276 77649-360
Cyclopentyl(3,4-dimethylphenyl)methanone ≥95%
Cyclopentyl(3,4-dimethylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₄H₁₈O
MW: 202.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03841315
CAS Number: 898791-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR